Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: GSL09 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001344150.1 | NP_001329981.1 | 36 - 37 | 4093 | 89 | |
NM_001344151.1 | NP_001329982.1 | 36 - 37 | 4093 | 89 | ||
NM_001344152.1 | NP_001329979.1 | 35 - 36 | 4164 | 89 | ||
NM_001344153.1 | NP_001329980.1 | 35 - 36 | 4010 | 89 | ||
NM_123045.4 | NP_198503.3 | 35 - 36 | 4013 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |