Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: TPS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001341861.1 | NP_001327998.1 | 5 - 6 | 860 | 147 | |
NM_118890.2 | NP_194485.1 | 5 - 6 | 860 | 147 | ||
GenBank mRNA | KJ676985.1 | - | 5 - 6 | 820 | 147 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |