Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIF7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037040.3 | NP_001032117.1 | 1 - 2 | 893 | 86 | |
NM_001345463.1 | NP_001331793.1 | 2 - 3 | 603 | 86 | ||
NM_125520.5 | NP_200935.2 | 2 - 3 | 749 | 86 | ||
GenBank mRNA | AB493805.1 | - | 1 - 2 | 423 | 86 | |
AJ630484.1 | - | 1 - 2 | 159 | 86 | ||
AY568656.1 | - | 1 - 2 | 159 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |