Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: FER2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111942.4 | NP_187716.1 | 1 - 2 | 465 | 76 | |
GenBank mRNA | AJ312192.1 | - | 1 - 2 | 324 | 76 | |
AK175807.1 | - | 1 - 2 | 345 | 76 | ||
AK175899.1 | - | 1 - 2 | 345 | 76 | ||
AK175901.1 | - | 1 - 2 | 345 | 76 | ||
AK175910.1 | - | 1 - 2 | 345 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |