Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111987.4 | NP_187761.1 | 8 - 9 | 1760 | 101 | |
GenBank mRNA | AK221314.1 | - | 8 - 9 | 1486 | 101 | |
KJ138828.1 | - | 7 - 8 | 1125 | 101 | ||
U62135.1 | - | 8 - 9 | 1475 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |