Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: HRS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_114479.5 | NP_190196.1 | 8 - 9 | 1095 | 62 | |
GenBank mRNA | AF020715.1 | - | 8 - 9 | 1031 | 62 | |
AY056233.1 | - | 8 - 9 | 1036 | 62 | ||
AY117252.1 | - | 8 - 9 | 1026 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |