Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATSPS4F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036532.3 | NP_001031609.1 | 12 - 13 | 2082 | 87 | |
NM_001340652.1 | NP_001329003.1 | 11 - 12 | 3023 | 87 | ||
NM_001340653.1 | NP_001329002.1 | 10 - 11 | 2311 | 87 | ||
NM_001340654.1 | NP_001329004.1 | 8 - 9 | 1341 | 87 | ||
NM_117080.5 | NP_192750.2 | 12 - 13 | 2932 | 87 | ||
GenBank mRNA | KJ138763.1 | - | 11 - 12 | 1904 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |