Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: OPT7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340684.1 | NP_001328829.1 | 5 - 6 | 1451 | 89 | |
NM_117145.3 | NP_192815.1 | 6 - 7 | 1412 | 89 | ||
GenBank mRNA | AY045668.1 | - | 6 - 7 | 1336 | 89 | |
BT004519.1 | - | 6 - 7 | 1253 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |