Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BPIFA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174426.3 | NP_776851.1 | 3 - 4 | 364 | 61 | |
GenBank mRNA | AF488706.1 | - | 3 - 4 | 365 | 61 | |
BC114803.1 | - | 3 - 4 | 377 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |