Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
Gene Symbol: MMACHC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015588.3 | NP_001015588.2 | 1 - 2 | 208 | 61 | |
GenBank mRNA | BC110137.1 | - | 1 - 2 | 209 | 61 | |
BT020992.1 | - | 1 - 2 | 85 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |