Search Thermo Fisher Scientific
Gene Symbol: GHDC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075703.1 | NP_001069171.1 | 4 - 5 | 578 | 81 | |
GenBank mRNA | BT026535.1 | - | 4 - 5 | 578 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Bos taurus Probe spans exons |