Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: SAV1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191362.2 | NP_001178291.1 | 1 - 2 | 472 | 122 | |
GenBank mRNA | BC123794.1 | - | 1 - 2 | 417 | 122 | |
EH152231.1 | - | 1 - 2 | 478 | 122 | ||
EH376871.1 | - | 1 - 2 | 448 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |