Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: MAP1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206119.3 | NP_001193048.2 | 6 - 7 | 7305 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Bos taurus Probe spans exons |