Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: prk-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001276777.3 | NP_001263706.1 | 6 - 7 | 855 | 99 | |
NM_001377564.1 | NP_001364690.1 | 5 - 6 | 598 | 99 | ||
NM_001377565.2 | NP_001364691.1 | 5 - 6 | 598 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |