Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: exc-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027868.5 | NP_001023039.1 | 5 - 6 | 633 | 123 | |
NM_001307099.3 | NP_001294028.1 | 4 - 5 | 432 | 123 | ||
NM_001307100.4 | NP_001294029.1 | 6 - 7 | 869 | 123 | ||
GenBank mRNA | AB060647.1 | - | 5 - 6 | 686 | 123 | |
AF339059.1 | - | 5 - 6 | 686 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Caenorhabditis elegans Probe spans exons |