Search
Search

unc-62| Gene Symbol: unc-62 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001028998.4 | NP_001024169.1 | 3 - 4 | 465 | 126 | |
| NM_001028999.5 | NP_001024170.1 | 3 - 4 | 816 | 126 | ||
| NM_001029002.4 | NP_001024173.1 | 2 - 3 | 350 | 126 | ||
| NM_001029004.5 | NP_001024175.1 | 2 - 3 | 239 | 126 | ||
| NM_001313278.3 | NP_001300207.1 | 2 | 239 | 126 | ||
| NM_001380634.1 | NP_001367924.1 | 3 - 4 | 465 | 126 | ||
| NM_001380635.1 | NP_001367923.1 | 3 - 4 | 467 | 126 | ||
| NM_001383298.1 | 3 - 4 | 459 | 126 | |||
| GenBank mRNA | AF427474.1 | - | 3 - 4 | 487 | 126 | |
| AF427475.1 | - | 3 - 4 | 487 | 126 | ||
| AF427476.1 | - | 3 - 4 | 670 | 126 | ||
| AF427477.1 | - | 3 - 4 | 670 | 126 | ||
Set Membership: |
|