Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: trr-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001377877.2 | NP_001364647.1 | 20 - 21 | 11535 | 80 | |
NM_001377878.3 | NP_001364646.1 | 20 - 21 | 11568 | 80 | ||
NM_001377879.1 | NP_001364592.1 | 20 - 21 | 11639 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |