Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: nfm-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027400.4 | NP_001022571.1 | 4 - 5 | 786 | 127 | |
NM_065934.5 | NP_498335.1 | 4 - 5 | 1212 | 127 | ||
NM_065935.5 | NP_498336.1 | 4 - 5 | 1176 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |