Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: glr-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_066486.8 | NP_498887.2 | 2 - 3 | 218 | 90 | |
GenBank mRNA | U34661.1 | - | 2 - 3 | 297 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |