Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mvb-12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_068901.7 | NP_501302.1 | 2 - 3 | 243 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |