Search Thermo Fisher Scientific
Gene Symbol: rom-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047548.5 | NP_001041013.2 | 9 - 10 | 1695 | 68 | |
NM_001083250.6 | NP_001076719.2 | 9 - 10 | 1701 | 68 | ||
NM_001380564.1 | NP_001368544.1 | 2 - 3 | 750 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Caenorhabditis elegans Probe spans exons |