Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: sulp-8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001269200.3 | NP_001256129.1 | 4 - 5 | 1471 | 91 | |
NM_001269201.3 | NP_001256130.1 | 2 - 3 | 1137 | 91 | ||
NR_069213.1 | 4 - 5 | 1472 | 91 | |||
GenBank mRNA | AY887917.1 | - | 4 - 5 | 1494 | 91 | |
AY954525.1 | - | 3 - 4 | 1392 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |