Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
Gene Symbol: CCL25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005259.1 | NP_001005259.1 | 2 - 3 | 183 | 66 | |
XM_005632314.3 | XP_005632371.1 | 3 - 4 | 292 | 66 | ||
XM_005632315.3 | XP_005632372.1 | 3 - 4 | 1563 | 66 | ||
XM_022406207.1 | XP_022261915.1 | 7 - 8 | 2999 | 66 | ||
XM_022406208.1 | XP_022261916.1 | 7 - 8 | 2543 | 66 | ||
XM_022406209.1 | XP_022261917.1 | 8 - 9 | 2374 | 66 | ||
XM_022406210.1 | XP_022261918.1 | 8 - 9 | 1852 | 66 | ||
XM_022406211.1 | XP_022261919.1 | 8 - 9 | 1855 | 66 | ||
XM_022406212.1 | XP_022261920.1 | 7 - 8 | 2226 | 66 | ||
XM_022406213.1 | XP_022261921.1 | 3 - 4 | 1836 | 66 | ||
XM_022406214.1 | XP_022261922.1 | 4 - 5 | 1212 | 66 | ||
XM_022406215.1 | XP_022261923.1 | 4 - 5 | 1646 | 66 | ||
GenBank mRNA | AB164617.1 | - | 2 - 3 | 183 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Canis familiaris Probe spans exons |