Search
Search

DNAJC22| Gene Symbol: DNAJC22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022411296.1 | XP_022267004.1 | 3 - 4 | 1095 | 123 | |
| XM_543683.6 | XP_543683.3 | 3 - 4 | 1677 | 123 | ||
Set Membership: |
|