Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003503109.2 | XP_003503157.1 | 4 - 5 | 515 | 60 | |
XM_007643735.1 | XP_007641925.1 | 4 - 5 | 518 | 60 | ||
XM_007643737.1 | XP_007641927.1 | 4 - 5 | 518 | 60 | ||
GenBank mRNA | JI889128.1 | - | 4 - 5 | 495 | 60 | |
JP047060.1 | - | 4 - 5 | 496 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Probe spans exons |