Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Rdh8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003499776.1 | XP_003499824.1 | 3 - 4 | 450 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Probe spans exons |