Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mogs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003511143.2 | XP_003511191.2 | 1 - 2 | 42 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Inventoried Probe spans exons |