Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Pptc7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007627804.1 | XP_007625994.1 | 4 - 5 | 701 | 89 | |
XM_007627811.1 | XP_007626001.1 | 4 - 5 | 524 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Protein Phosphatase Probe spans exons |