Search Thermo Fisher Scientific
Gene Symbol: Smg8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003503805.2 | XP_003503853.1 | 1 - 2 | 1767 | 105 | |
GenBank mRNA | JP045092.1 | - | 1 - 2 | 988 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Probe spans exons |