Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: VPS35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005691946.3 | XP_005692003.1 | 11 - 12 | 1450 | 139 | |
GenBank mRNA | JO426407.1 | - | 6 - 7 | 1725 | 139 |
Set Membership: |
Amplicon length greater than or equal to 101 Capra hircus Probe spans exons |