Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APOA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005689482.3 | XP_005689539.3 | 1 - 2 | 318 | 59 | |
GenBank mRNA | JO420626.2 | - | 2 - 3 | 1258 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons |