Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: BEND2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018043708.1 | XP_017899197.1 | 13 - 14 | 2015 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |