Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: SLC22A11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005699974.2 | XP_005700031.2 | 3 - 4 | 651 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Capra hircus Probe spans exons |