Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Ge-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_135642.3 | NP_609486.2 | 10 - 11 | 4128 | 88 | |
NM_164957.2 | NP_723654.1 | 9 - 10 | 4211 | 88 | ||
GenBank mRNA | BT004479.1 | - | 10 - 11 | 4108 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Drosophila melanogaster |