Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Cul1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078931.4 | NP_523655.1 | 2 - 3 | 1237 | 91 | |
NM_165569.2 | NP_724621.1 | 2 - 3 | 1260 | 91 | ||
NM_165570.3 | NP_724622.1 | 2 - 3 | 1258 | 91 | ||
NM_165571.2 | NP_724623.1 | 3 - 4 | 1278 | 91 | ||
GenBank mRNA | AF136343.1 | - | 2 - 3 | 1285 | 91 | |
BT010290.1 | - | 2 - 3 | 1277 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |