Search
Search

rdgB| Gene Symbol: rdgB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014740.2 | NP_001014740.1 | 7 - 8 | 1608 | 93 | |
| NM_001014741.3 | NP_001014741.1 | 8 - 9 | 1666 | 93 | ||
| NM_001169277.2 | NP_001162748.1 | 7 - 8 | 1608 | 93 | ||
| NM_001169278.3 | NP_001162749.1 | 7 - 8 | 1608 | 93 | ||
| NM_001169279.3 | NP_001162750.1 | 7 - 8 | 1608 | 93 | ||
| NM_001169280.3 | NP_001162751.1 | 8 - 9 | 1647 | 93 | ||
| NM_001272605.2 | NP_001259534.1 | 9 - 10 | 1728 | 93 | ||
| NM_001272606.1 | NP_001259535.1 | 8 - 9 | 1762 | 93 | ||
| NM_001298300.1 | NP_001285229.1 | 7 - 8 | 1608 | 93 | ||
| NM_078594.3 | NP_511149.2 | 6 - 7 | 1652 | 93 | ||
| NM_167382.3 | NP_727733.1 | 7 - 8 | 1646 | 93 | ||
| GenBank mRNA | AY051422.1 | - | 7 - 8 | 1628 | 93 | |
Set Membership: |
|