Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
Gene Symbol: Or13a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078635.3 | NP_523359.2 | 4 - 5 | 982 | 99 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor Drosophila melanogaster |