Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Tsp42Ee | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273824.1 | NP_001260753.1 | 2 - 3 | 153 | 83 | |
NM_078907.3 | NP_523631.1 | 2 - 3 | 166 | 83 | ||
GenBank mRNA | AF220043.1 | - | 2 - 3 | 134 | 83 | |
BT044107.1 | - | 2 - 3 | 164 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Drosophila melanogaster |