Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: cno | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275342.1 | NP_001262271.1 | 5 - 6 | 1138 | 60 | |
NM_001275344.1 | NP_001262273.1 | 10 - 11 | 2418 | 60 | ||
NM_001275345.1 | NP_001262274.1 | 10 - 11 | 2418 | 60 | ||
NM_001316565.1 | NP_001303494.1 | 10 - 11 | 2418 | 60 | ||
NM_079508.4 | NP_524232.2 | 10 - 11 | 2418 | 60 | ||
NM_169026.3 | NP_730892.2 | 10 - 11 | 2418 | 60 | ||
GenBank mRNA | BT044227.1 | - | 10 - 11 | 2387 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |