Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: scu | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001258800.2 | NP_001245729.1 | 3 | 1025 | 97 | |
NM_078672.5 | NP_523396.1 | 2 | 925 | 97 | ||
GenBank mRNA | BT029045.1 | - | 2263 | 97 | ||
BT122199.1 | - | 3 | 956 | 97 | ||
Y15102.1 | - | 1841 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Drosophila melanogaster |