Search Thermo Fisher Scientific
Gene Symbol: trpn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_183349.1 | NP_899192.1 | 24 - 25 | 4866 | 75 | |
GenBank mRNA | AY313897.1 | - | 24 - 25 | 4866 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Danio rerio Probe spans exons |