Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
Gene Symbol: dixdc1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_182884.1 | NP_878304.1 | 12 - 13 | 1492 | 75 | |
GenBank mRNA | AF502143.1 | - | 12 - 13 | 1492 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Danio rerio Probe spans exons |