Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tmcc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044919.1 | NP_001038384.1 | 3 - 4 | 1155 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Danio rerio Probe spans exons |