Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hapln3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213101.1 | NP_998266.1 | 2 - 3 | 220 | 130 | |
GenBank mRNA | BC068325.1 | - | 2 - 3 | 220 | 130 | |
CU458908.1 | - | 2 - 3 | 220 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |