Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFT46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015298265.1 | XP_015153751.1 | 8 - 9 | 1013 | 65 | |
XM_417918.6 | XP_417918.5 | 8 - 9 | 1016 | 65 | ||
GenBank mRNA | CR406386.1 | - | 6 - 7 | 546 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Gallus gallus Probe spans exons |