Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: PARPBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030763.2 | NP_001025934.2 | 9 - 10 | 1358 | 149 | |
XM_025148736.1 | XP_025004504.1 | 9 - 10 | 1351 | 149 | ||
XM_025148737.1 | XP_025004505.1 | 9 - 10 | 1361 | 149 | ||
GenBank mRNA | AJ720065.1 | - | 9 - 10 | 1358 | 149 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |