Search
Search

BLEC2| Gene Symbol: BLEC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044682.1 | NP_001038147.1 | 1 | 52 | 53 | |
| GenBank mRNA | AJ245903.1 | - | 1 | 65 | 53 | |
| AJ634338.1 | - | 1 | 52 | 53 | ||
| MF563587.1 | - | 1 | 52 | 53 | ||
Set Membership: |
|