Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00153451_m1
          See other E2F1 GE Assays ›
          Gene Symbol:
          E2F1
          Gene Name:
          E2F transcription factor 1
          Gene Aliases:
          E2F-1, RBAP1, RBBP3, RBP3
          Chromosome Location:
          Chr.20: 33675477 - 33686385 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00153451_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: E2F1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_005225.3 NP_005216.1 3 - 4 696 84
          XM_047439961.1 XP_047295917.1 3 - 4 696 84
          GenBank mRNA BC050369.2 - 3 - 4 715 84
          BC058902.1 - 3 - 4 646 84
          M96577.1 - 3 - 4 710 84
          S49592.1 - 3 - 4 692 84

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC2000008894.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC20000773.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Homo sapiens Human Hedgehog Pathway Human Transcription Factors (non-Hox) Human p53 Signaling HUMAN CYCLINS AND CELL CYCLE REGULATION HUMAN MOLECULAR MECHANISMS OF CANCER HUMAN BRCA1 PATHWAY HUMAN BREAST CANCER REGULATION BY STATHMIN1 HUMAN FACTORS PROMOTING CARDIOGENESIS IN VERTEBRATES HUMAN PI3K SIGNALING HUMAN PANCREATIC ADENOCARCINOMA Inventoried Human_Hedgehog Human_p53_Signaling Human_Transcription_Factors_(non-Hox) Human_Pancreatic_Adenocarcinoma Human_Factors_Promoting_Cardiogenesis_in_Vertebrates Human_BRCA1_Pathway Human_Molecular_Mechanisms_of_Cancer Human_Cyclins_and_Cell_Cycle_Regulation Human_PI3K_Signaling Human_Breast_Cancer_Regulation_by_Stathmin1 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene E2F1

          Gene Ontology Categories:

          Function(s) Process(es)

          DNA damage checkpoint
          negative regulation of transcription from RNA polymerase II promoter
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          regulation of transcription from RNA polymerase II promoter
          apoptotic process
          spermatogenesis
          intrinsic apoptotic signaling pathway in response to DNA damage
          positive regulation of gene expression
          forebrain development
          response to lipopolysaccharide
          positive regulation of apoptotic process
          anoikis
          negative regulation of DNA binding
          negative regulation of fat cell differentiation
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of fibroblast proliferation
          mRNA stabilization
          regulation of cell cycle
          positive regulation of glial cell proliferation
          negative regulation of fat cell proliferation
          cellular response to fatty acid
          cellular response to hypoxia
          cellular response to xenobiotic stimulus
          intrinsic apoptotic signaling pathway by p53 class mediator
          lens fiber cell apoptotic process
          cellular response to nerve growth factor stimulus
          regulation of G1/S transition of mitotic cell cycle
          transcription regulatory region sequence-specific DNA binding
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          RNA polymerase II transcription factor activity, sequence-specific DNA binding
          core promoter proximal region sequence-specific DNA binding
          bacterial-type RNA polymerase transcriptional activator activity, sequence-specific DNA binding
          DNA binding
          transcription factor activity, sequence-specific DNA binding
          protein binding
          protein kinase binding
          sequence-specific DNA binding
          protein dimerization activity
          binding, bridging
          DNA-binding transcription factor binding
          sequence-specific double-stranded DNA binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC2000008894.hg.1 ENST00000343380.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000008894.hg.1 ENST00000343380.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000008894.hg.1 ENST00000343380.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000008894.hg.1 ENST00000343380.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000008894.hg.1 ENST00000343380.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000008894.hg.1 ENST00000343380.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000008894.hg.1 ENST00000343380.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000008894.hg.1 ENST00000343380.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000008894.hg.1 ENST00000343380.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000008894.hg.1 ENST00000343380.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000008894.hg.1 NM_005225.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000008894.hg.1 NM_005225.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000008894.hg.1 NM_005225.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000008894.hg.1 NM_005225.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000008894.hg.1 NM_005225.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000008894.hg.1 NM_005225.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000008894.hg.1 NM_005225.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000008894.hg.1 NM_005225.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000008894.hg.1 NM_005225.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000008894.hg.1 NM_005225.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000008894.hg.1 NM_005225.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR20010353.hg.1 ENST00000343380.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR20010353.hg.1 NM_005225.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR20010353.hg.1 TC20000773.hg.1 HTA TC
          Affymetrix Human Clariom D Assay TC2000008894.hg.1 ENST00000343380.5 Ensembl
          Affymetrix Human Clariom D Assay TC2000008894.hg.1 ENST00000343380.5 Ensembl
          Affymetrix Human Clariom D Assay TC2000008894.hg.1 ENST00000343380.5 Ensembl
          Affymetrix Human Clariom D Assay TC2000008894.hg.1 ENST00000343380.5 Ensembl
          Affymetrix Human Clariom D Assay TC2000008894.hg.1 ENST00000343380.5 Ensembl
          Affymetrix Human Clariom D Assay TC2000008894.hg.1 ENST00000343380.5 Ensembl
          Affymetrix Human Clariom D Assay TC2000008894.hg.1 ENST00000343380.5 Ensembl
          Affymetrix Human Clariom D Assay TC2000008894.hg.1 ENST00000343380.5 Ensembl
          Affymetrix Human Clariom D Assay TC2000008894.hg.1 ENST00000343380.5 Ensembl
          Affymetrix Human Clariom D Assay TC2000008894.hg.1 ENST00000343380.5 Ensembl
          Affymetrix Human Clariom D Assay TC2000008894.hg.1 NM_005225.2 Refseq
          Affymetrix Human Clariom D Assay TC2000008894.hg.1 NM_005225.2 Refseq
          Affymetrix Human Clariom D Assay TC2000008894.hg.1 NM_005225.2 Refseq
          Affymetrix Human Clariom D Assay TC2000008894.hg.1 NM_005225.2 Refseq
          Affymetrix Human Clariom D Assay TC2000008894.hg.1 NM_005225.2 Refseq
          Affymetrix Human Clariom D Assay TC2000008894.hg.1 NM_005225.2 Refseq
          Affymetrix Human Clariom D Assay TC2000008894.hg.1 NM_005225.2 Refseq
          Affymetrix Human Clariom D Assay TC2000008894.hg.1 NM_005225.2 Refseq
          Affymetrix Human Clariom D Assay TC2000008894.hg.1 NM_005225.2 Refseq
          Affymetrix Human Clariom D Assay TC2000008894.hg.1 NM_005225.2 Refseq
          Affymetrix Human Clariom D Assay TC2000008894.hg.1 NM_005225.2 Refseq
          Affymetrix Human Clariom D Assay PSR2000161989.hg.1 ENST00000343380.5 Ensembl
          Affymetrix Human Clariom D Assay PSR2000161989.hg.1 NM_005225.2 Refseq
          Affymetrix Human Clariom D Assay PSR2000161989.hg.1 ENST00000343380.5 Ensembl
          Affymetrix Human Clariom D Assay PSR2000161989.hg.1 NM_005225.2 Refseq
          Affymetrix Human Clariom D Assay PSR2000161989.hg.1 TC2000008894.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline