Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: CNOT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001265612.1 | NP_001252541.1 | 46 - 47 | 7099 | 143 | |
NM_016284.4 | NP_057368.3 | 46 - 47 | 7114 | 143 | ||
NR_049763.1 | 47 - 48 | 7282 | 143 | |||
GenBank mRNA | AB023224.2 | - | 33 - 34 | 5159 | 143 | |
AF110778.1 | - | 6 - 7 | 891 | 142 | ||
AK056927.1 | - | 23 - 24 | 3444 | 143 | ||
AK075125.1 | - | 16 - 17 | 2413 | 143 | ||
AK092885.1 | - | 6 - 7 | 1658 | 143 | ||
AK301092.1 | - | 6 - 7 | 878 | 143 | ||
AK302495.1 | - | 23 - 24 | 3484 | 143 | ||
AL117492.1 | - | 18 - 19 | 2953 | 143 | ||
AL833549.1 | - | 46 - 47 | 7073 | 143 | ||
BC000779.2 | - | 8 - 9 | 1272 | 143 | ||
BC024317.1 | - | 29 - 30 | 4513 | 143 | ||
BC027969.2 | - | 5 - 6 | 728 | 143 | ||
BX537840.1 | - | 47 - 48 | 7228 | 143 | ||
EF553522.1 | - | 46 - 47 | 7048 | 143 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Inventoried Probe spans exons |