Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016432.hg.1 |
ENST00000367716.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016432.hg.1 |
ENST00000367716.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016432.hg.1 |
ENST00000367716.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016432.hg.1 |
ENST00000367716.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016432.hg.1 |
ENST00000367716.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016432.hg.1 |
ENST00000367716.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016432.hg.1 |
ENST00000367716.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016432.hg.1 |
ENST00000367716.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016432.hg.1 |
ENST00000367716.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016432.hg.1 |
ENST00000367716.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016432.hg.1 |
ENST00000367716.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016432.hg.1 |
ENST00000437190.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016432.hg.1 |
ENST00000437190.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016432.hg.1 |
ENST00000437190.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016432.hg.1 |
ENST00000437190.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016432.hg.1 |
ENST00000437190.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016432.hg.1 |
ENST00000437190.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016432.hg.1 |
NR_037845.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016432.hg.1 |
NR_037845.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016432.hg.1 |
NR_037845.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016432.hg.1 |
NR_037845.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016432.hg.1 |
NR_125960.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016432.hg.1 |
NR_125960.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016432.hg.1 |
NR_125960.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016432.hg.1 |
NR_125960.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016432.hg.1 |
NR_125960.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016432.hg.1 |
NR_125960.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016432.hg.1 |
NR_125960.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016432.hg.1 |
NR_125960.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016432.hg.1 |
NR_125960.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016432.hg.1 |
NR_125960.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016432.hg.1 |
NR_125960.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013914.hg.1 |
ENST00000464152.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013914.hg.1 |
ENST00000464152.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013914.hg.1 |
ENST00000464152.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013914.hg.1 |
ENST00000464152.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013914.hg.1 |
ENST00000473790.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013914.hg.1 |
ENST00000473790.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013914.hg.1 |
ENST00000473790.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013914.hg.1 |
ENST00000473790.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013914.hg.1 |
ENST00000479720.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013914.hg.1 |
ENST00000479720.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013914.hg.1 |
ENST00000479720.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013914.hg.1 |
ENST00000479720.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013914.hg.1 |
ENST00000488841.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013914.hg.1 |
ENST00000488841.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013914.hg.1 |
ENST00000488841.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013914.hg.1 |
ENST00000488841.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013914.hg.1 |
ENST00000488841.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013914.hg.1 |
ENST00000488841.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013914.hg.1 |
ENST00000488841.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013914.hg.1 |
ENST00000497628.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013914.hg.1 |
ENST00000497628.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013914.hg.1 |
ENST00000497628.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013914.hg.1 |
ENST00000497628.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013913.hg.1 |
ENST00000496671.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013913.hg.1 |
ENST00000496671.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013913.hg.1 |
ENST00000496671.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013913.hg.1 |
ENST00000496671.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016432.hg.1 |
ENST00000367716.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016432.hg.1 |
ENST00000367716.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016432.hg.1 |
ENST00000367716.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016432.hg.1 |
ENST00000367716.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016432.hg.1 |
ENST00000367716.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016432.hg.1 |
ENST00000367716.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016432.hg.1 |
ENST00000367716.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016432.hg.1 |
ENST00000367716.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016432.hg.1 |
ENST00000367716.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016432.hg.1 |
ENST00000367716.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016432.hg.1 |
ENST00000367716.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016432.hg.1 |
ENST00000437190.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016432.hg.1 |
ENST00000437190.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016432.hg.1 |
ENST00000437190.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016432.hg.1 |
ENST00000437190.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016432.hg.1 |
ENST00000437190.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016432.hg.1 |
ENST00000437190.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016432.hg.1 |
NR_037845.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016432.hg.1 |
NR_037845.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016432.hg.1 |
NR_037845.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016432.hg.1 |
NR_037845.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016432.hg.1 |
NR_125960.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016432.hg.1 |
NR_125960.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016432.hg.1 |
NR_125960.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016432.hg.1 |
NR_125960.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016432.hg.1 |
NR_125960.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016432.hg.1 |
NR_125960.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016432.hg.1 |
NR_125960.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016432.hg.1 |
NR_125960.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016432.hg.1 |
NR_125960.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016432.hg.1 |
NR_125960.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016432.hg.1 |
NR_125960.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013913.hg.1 |
ENST00000496671.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013913.hg.1 |
ENST00000496671.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013913.hg.1 |
ENST00000496671.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013913.hg.1 |
ENST00000496671.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013914.hg.1 |
ENST00000464152.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013914.hg.1 |
ENST00000464152.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013914.hg.1 |
ENST00000464152.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013914.hg.1 |
ENST00000464152.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013914.hg.1 |
ENST00000473790.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013914.hg.1 |
ENST00000473790.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013914.hg.1 |
ENST00000473790.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013914.hg.1 |
ENST00000473790.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013914.hg.1 |
ENST00000479720.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013914.hg.1 |
ENST00000479720.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013914.hg.1 |
ENST00000479720.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013914.hg.1 |
ENST00000479720.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013914.hg.1 |
ENST00000488841.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013914.hg.1 |
ENST00000488841.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013914.hg.1 |
ENST00000488841.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013914.hg.1 |
ENST00000488841.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013914.hg.1 |
ENST00000488841.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013914.hg.1 |
ENST00000488841.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013914.hg.1 |
ENST00000488841.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013914.hg.1 |
ENST00000497628.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013914.hg.1 |
ENST00000497628.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013914.hg.1 |
ENST00000497628.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013914.hg.1 |
ENST00000497628.1 |
Ensembl |